Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eps8 Antibody (18F1F2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP227300C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227300C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227300C Supplier Novus Biologicals Supplier No. NBP227300C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Eps8 Monoclonal specifically detects Eps8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Eps8
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 18F1F2
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Accession No. Q12929
Gene Alias epidermal growth factor receptor kinase substrate 8, epidermal growth factor receptor pathway substrate 8
Gene Symbols EPS8
Host Species Mouse
Immunogen Amino acids 701-882 of human EPS8 protein were used as the immunogen for the EPS8 antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2059
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.