Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPS8L2 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB181754 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181754 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181754 Supplier Novus Biologicals Supplier No. NBP306508H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EPS8L2 Polyclonal antibody specifically detects EPS8L2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen EPS8L2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias epidermal growth factor receptor kinase substrate 8-like protein 2, Epidermal growth factor receptor pathway substrate 8-related protein 2, EPS8-like 2, EPS8-like protein 2, EPS8R2, EPS8-related protein 2, FLJ16738, FLJ21935, FLJ22171, MGC126530, MGC3088
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human EPS8L2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64787
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.