Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPS8L3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB051783 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB051783 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB051783 Supplier Novus Biologicals Supplier No. NBP298054JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPS8L3 Polyclonal specifically detects EPS8L3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EPS8L3
Applications Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias epidermal growth factor receptor kinase substrate 8-like protein 3, Epidermal growth factor receptor pathway substrate 8-related protein 3, EPS8-like 3, EPS8-like protein 3, EPS8R3, EPS8-related protein 3, FLJ21522, MGC16817
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human EPS8L3 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79574
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.