Learn More
epsilon-Caprolactam, 99%
Description
ε-Caprolactam is used as a monomer in the preparation of synthetic fibers of the polyamide as well as a solvent for high molecular weight polymers. It is also used in coatings and plasticizers. Further, it is used as a cross linking agent for polyurethanes. In addition to this, it is used to prepare nylon-6, which finds application in fibers and plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ε-Caprolactam is used as a monomer in the preparation of synthetic fibers of the polyamide as well as a solvent for high molecular weight polymers. It is also used in coatings and plasticizers. Further, it is used as a cross linking agent for polyurethanes. In addition to this, it is used to prepare nylon-6, which finds application in fibers and plastics.
Solubility
Soluble in benzene, ethanol, water, chloroform, methanol, tetrahydrofurfuryl alcohol, ether, dimethylformamide, chlorinated hydrocarbons, cyclohexene and petroleum fractions.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 68°C to 72°C |
| Density | 1.01 |
| pH | 7 to 8.5 |
| Boiling Point | 270°C |
| Flash Point | 152°C (305°F) |
| Quantity | 2500 g |
| Beilstein | 106934 |
| Merck Index | 14,1761 |
| Solubility Information | Soluble in benzene,ethanol,water,chloroform,methanol,tetrahydrofurfuryl alcohol,ether,dimethylformamide,chlorinated hydrocarbons,cyclohexene and petroleum fractions. |
| Formula Weight | 113.16 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H317-H319-H335-H500 |
| Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c |
| EINECSNumber | 203-313-2 |
| RTECSNumber | CM3675000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.