Learn More
epsilon-Caprolactone, 99%
Description
ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.
Solubility
Miscible with water and most organic solvents.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Combustible liquid.

Specifications
Specifications
Melting Point | -2°C |
Density | 1.078 |
Boiling Point | 235°C to 236°C |
Flash Point | 109°C (228°F) |
Assay Percent Range | 99% |
Refractive Index | 1.463 |
Quantity | 2500 g |
Beilstein | 106919 |
Solubility Information | Miscible with water and most organic solvents. |
Formula Weight | 114.14 |
Show More |
Safety and Handling
Hazard Category | H319 |
Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
EINECSNumber | 207-938-1 |
RTECSNumber | MO8400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.