Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

epsilon-Caprolactone, 99%

Catalog No. p-7080670
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g

CAS: 502-44-3 | C6H10O2 | 114.144 g/mol

ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.

Solubility
Miscible with water and most organic solvents.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Combustible liquid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-44-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00003267
InChI Key PAPBSGBWRJIAAV-UHFFFAOYSA-N
Synonym 6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane
PubChem CID 10401
ChEBI CHEBI:17915
IUPAC Name oxepan-2-one
SMILES C1CCC(=O)OCC1

Specifications

Melting Point -2°C
Density 1.078
Boiling Point 235°C to 236°C
Flash Point 109°C (228°F)
Assay Percent Range 99%
Refractive Index 1.463
Quantity 100 g
Beilstein 106919
Solubility Information Miscible with water and most organic solvents.
Formula Weight 114.14
Percent Purity 99%
Chemical Name or Material epsilon-Caprolactone
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 207-938-1
RTECSNumber MO8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.