Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epstein Barr Virus Antibody (15F9), HRP, Novus Biologicals™
SDP

Catalog No. NB10066574H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10066574H 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10066574H Supplier Novus Biologicals Supplier No. NB10066574H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Epstein Barr Virus Monoclonal specifically detects Epstein Barr Virus in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Epstein Barr Virus
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 15F9
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Alias Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1
Host Species Rat
Immunogen Bacterial TrpE-LMP2A fusion protein
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes latent membrane protein 2A (LMP2A) of Epstein-Barr virus (EBV). EBV is a human herpesvirus, which is associated with conditions such as Hodgkin
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.