Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epstein Barr Virus Antibody (CS1-4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB392169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392169 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392169 Supplier Novus Biologicals Supplier No. NBP308501AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Epstein Barr Virus Monoclonal antibody specifically detects Epstein Barr Virus in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Epstein Barr Virus
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CS1-4
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1
Host Species Mouse
Immunogen Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.