Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epstein Barr Virus Nuclear Antigen 4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP266590 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266590 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266590 Supplier Novus Biologicals Supplier No. NBP266590
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Epstein Barr Virus Nuclear Antigen 4 Polyclonal specifically detects Epstein Barr Virus Nuclear Antigen 4 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot.

Specifications

Antigen EpsteinBarrVirusNuclearAntigen4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml
Formulation Delipidated, heat inactivated, lyophilized stable whole serum with No Preservative
Gene Alias Epstein Barr Virus Nuclear Antigen 3B
Host Species Sheep
Immunogen Recombinant EBNA-3B protein
Purification Method Whole Antiserum
Quantity 250 μg
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.