Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPX Antibody (EPX/3908R), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB392223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392223 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392223 Supplier Novus Biologicals Supplier No. NBP308476AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EPX Monoclonal antibody specifically detects EPX in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen EPX
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EPX/3908R
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias EC 1.11.1.7, eosinophil peroxidase, EPOEPER, EPPEC 1.11.1, EPX-PEN
Host Species Rabbit
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome (Uniprot: P11678 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8288
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.