Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Equilateral Prisms
SCI_ED

Catalog No. p-7145471 Shop All Eisco Products
Click to view available options
Length (Metric):
25 mm
50 mm
75 mm
100 mm
150 mm

Catalog No. S41530

Supplier: Eisco™ PH0554HG

Only null left
Add to Cart
Add to Cart

These equilateral prisms are polished on all faces. Designed for use in optical experiments. Prisms will create a nice rainbow.

  • Made with optical quality glass or clear acrylic
  • Polished on all faces
  • Designed for use in optical experiments
  • Will create a nice rainbow

Warranty and Services

1 year limited manufacturer warranty

Specifications

Product Type Equilateral Solid
Material Glass
For Use With (Application) Optics experiments and demonstrations
Quantity 1
Length (Metric) 50 mm
Weight (Metric) 36 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.