Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ER 50891
GREENER_CHOICESDP

Catalog No. 382310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-231-0 10 mg
38-235-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-231-0 Supplier Tocris Bioscience™ Supplier No. 3823/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective RARα antagonist

Antagonist of RARα receptors (IC50 = 31.2 nM). Displays selective affinity for RARα (relative IC50 values are 1.8, 432 and 535 nM for RARα, RARγ and RARβ respectively).

Chemical Identifiers

CAS 187400-85-7
Molecular Formula C29H24N2O2

Specifications

Quantity 10 mg
Formula Weight 432.51
Purity >99%
Molecular Weight (g/mol) 432.523
Chemical Name or Material ER 50891
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.