Learn More
Description
Specifications
Specifications
| Antigen | ER alpha/NR3A1 |
| Applications | ChIP Assay, ELISA, Gel Shift, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | C.15100066 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:5000, Chromatin Immunoprecipitation 2.5 uL/IP, ELISA 1:500 - 1:5000, Immunohistochemistry 1:200 - 1:5000, Immunoprecipitation 1:200 - 1:5000, Gel Supershift Assay 1:10 - 1:20, Chromatin Immunoprecipitation Sequencing 2.5 uL/IP |
| Formulation | Ascites Fluid with 0.05% Sodium Azide |
| Gene Alias | ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 |
| Gene Symbols | ESR1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
