Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER alpha/NR3A1 Antibody (C-542), Novus Biologicals™
SDP

Catalog No. NBP119318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119318 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119318 Supplier Novus Biologicals Supplier No. NBP119318
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen ER alpha/NR3A1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C-542
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Gene Accession No. P03372
Gene Alias ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1
Gene Symbols ESR1
Host Species Mouse
Immunogen Synthetic peptide corresponding to the sequence near the C-terminus of human Estrogen Receptor alpha.
Molecular Weight of Antigen 66 kDa
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2099
Test Specificity Estrogen Receptor alpha (C-542)
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.