Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER alpha/NR3A1 Mouse, Clone: AER314, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | ER alpha/NR3A1 |
---|---|
Clone | AER314 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP248012B
![]() |
Novus Biologicals
NBP2480120.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
ER alpha/NR3A1 Monoclonal antibody specifically detects Antigen in Bovine, Human, Mouse, Rabbit, Rat samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
ER alpha/NR3A1 | |
1.0 mg/mL | |
Flow Cytometry, Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
PBS with No Preservative | |
2099 | |
ER purified from cow uterus | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
AER314 | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine, Rabbit | |
ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
ESR1 | |
IgG1 | |
Protein A or G purified | |
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa120-170. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title