Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER alpha/NR3A1 Rabbit anti-Human, Clone: IHC423, Novus Biologicals™
SDP

Catalog No. NB120514 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120514 0.1 mL
NB413407 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB120514 Supplier Novus Biologicals Supplier No. NBP3117120.1ML
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ER alpha/NR3A1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IHC423
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200
Formulation Tris Buffer (pH 7.6) with BSA
Gene Alias ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2099
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.