Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER alpha/NR3A1 Rabbit anti-Human, Clone: IHC423, Novus Biologicals™

Rabbit Monoclonal Antibody
$219.50 - $939.00
Specifications
Antigen | ER alpha/NR3A1 |
---|---|
Clone | IHC423 |
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ER alpha/NR3A1 | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 | |
Unconjugated | |
Rabbit | |
Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
Tris Buffer (pH 7.6) with BSA | |
2099 | |
IgG κ | |
Affinity purified |
IHC423 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
Recombinant Protein | |
Primary | |
Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title