Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER beta/NR3A2 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB030117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB030117 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB030117 Supplier Novus Biologicals Supplier No. NBP281132DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ER beta/NR3A2 Polyclonal specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ER beta/NR3A2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2
Host Species Rabbit
Immunogen Partial recombinant human ER beta/NR3A2 protein (amino acids 1-153) [UniProt Q92731]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2100
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.