Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER-HR3 Antibody (ER-HR3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017667 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017667 0.025 mg
NB10064931 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017667 Supplier Novus Biologicals Supplier No. NB100649310.025MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

ER-HR3 Monoclonal specifically detects ER-HR3 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen ER-HR3
Applications Immunohistochemistry (Frozen), Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Flow Cytometry
Classification Monoclonal
Clone ER-HR3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Alias Hematopoiesis related macrophage
Host Species Rat
Immunogen Adherent F1 (CBAxBL) bone marrow stormal cells.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64931 recognizes the murine antigen ER-HR3. ER-HR3 is a cell surface antigen which is expressed by Langerhans cells in epithelium, a subset of mature macrophages and dendritic cells that are located predominantly in haematopoietic and lymphoid organs. Initial studies of ER-HR3 demonstrated expression on peripheral blood monocytes but recent studies suggest that expression is actually very low. During foetal development, ER-HR3 positive cells are localised to haemapoietic islands and are often associated with erythroid progenitor cells. The functions of the ER-HR3 antigen have not been established but reports suggest that the antigen may be involved in adult erythropoiesis and in the regulation of the immune response. Clone ER-HR3 does not inhibit T cell proliferation in antigen-specific T-cell proliferation studies. Clone ER-HR3 recognizes two proteins of approximately 69kD and 55kD under non-reducing conditions.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2c
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.