Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERAB Rabbit anti-Human, Mouse, Rat, Clone: 3O4W10, Novus Biologicals™
SDP

Catalog No. NB165750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165750 100 μg
NB165751 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165750 Supplier Novus Biologicals Supplier No. NBP315327100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ERAB Monoclonal antibody specifically detects ERAB in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ERAB
Applications Western Blot
Classification Monoclonal
Clone 3O4W10
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 17-beta-HSD 10, 17-beta-hydroxysteroid dehydrogenase 10, 17b-HSD10, 3-hydroxy-2-methylbutyryl-CoA dehydrogenase, ABAD, amyloid-beta peptide binding alcohol dehydrogenase, CAMR, DUPXp11.22, EC 1.1.1.178, EC 1.1.1.35, Endoplasmic reticulum-associated amyloid beta-peptide-binding protein, ERAB3-hydroxyacyl-CoA dehydrogenase type-2, HADH2AB-binding alcohol dehydrogenase, hydroxyacyl-Coenzyme A dehydrogenase, type II, hydroxyacyl-Coenzyme Adehydrogenase, type II, hydroxysteroid (17-beta) dehydrogenase 10, mental retardation, X-linked, syndromic 10, MHBD, Mitochondrial ribonuclease P protein 2,3-hydroxyacyl-CoA dehydrogenase type II, Mitochondrial RNase P protein 2, MRPP2HCD2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1, short chain dehydrogenase/reductase family 5C, member 1, short chain L-3-hydroxyacyl-CoA dehydrogenase type 2, short chain type dehydrogenase/reductase XH98G2, Short-chain type dehydrogenase/reductase XH98G2, Type II HADH, XH98G2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 162-261 of human ERAB (Q99714).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3028
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.