Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERAS Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB325526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325526 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325526 Supplier Novus Biologicals Supplier No. NBP299426PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERAS Polyclonal antibody specifically detects ERAS in Mouse samples. It is validated for Western Blot

Specifications

Antigen ERAS
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias Embryonic stem cell-expressed Ras, E-Ras, ES cell expressed Ras, HRAS2MGC126693, HRASPGTPase ERas, MGC126691, small GTPase protein E-Ras, v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2, v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ERAS
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3266
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.