Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB3/Her3 Antibody (138), Alexa Fluor™ 350
SDP

Catalog No. NB077641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB077641 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB077641 Supplier Novus Biologicals Supplier No. NBP290769AF350

Rabbit Monoclonal Antibody

ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen ErbB3/Her3
Applications ELISA
Classification Monoclonal
Clone 138
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ErbB3/Her3 (Q61526; Met1-His641).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2065
Test Specificity Has cross-reactivity in ELISA with: Human ERBB3
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.