Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB3/Her3 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP232253IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232253IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232253IR Supplier Novus Biologicals Supplier No. NBP232253IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ErbB3/Her3 Polyclonal specifically detects ErbB3/Her3 in Human samples. It is validated for Western Blot.

Specifications

Antigen ErbB3/Her3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Gene Symbols ERBB3
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1292 to 1342 of human V-Erb-B2 Erythroblastic Leukemia Viral Oncogene Homolog 3 using the numbering given in entry NP_001973.2 (GeneID 2065).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2065
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.