Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB3/Her3 Antibody (OTI3E10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270661V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270661V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270661V Supplier Novus Biologicals Supplier No. NBP270661V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen ErbB3/Her3
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3E10
Conjugate DyLight 405
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Gene Symbols ERBB3
Host Species Mouse
Immunogen Full length recombinant protein of human ErbB3/Her3 (NP_001973) produced in SF9 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2065.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.