Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB3/Her3 Antibody (SGP1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB1002692IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002692IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002692IR Supplier Novus Biologicals Supplier No. NB1002692IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen ErbB3/Her3
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone SGP1
Conjugate DyLight 755
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Gene Symbols ERBB3
Host Species Mouse
Immunogen c-erb B3 protein from transfected human kidney fibroblasts purified by wheatgerm lectin affinity chromatography
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2065
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.