Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB4/Her4 Antibody (HFR-1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119628 Supplier Novus Biologicals Supplier No. NBP308902
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ErbB4/Her4
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone HFR-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian)
Host Species Mouse
Immunogen Recombinant human ErbB4/Her4 protein fragment (around aa 1116-1269) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2066
Target Species Human, Mouse
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.