Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERBB4 Monoclonal Antibody (OTI1H6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
$516.29
Specifications
Antigen | ERBB4 |
---|---|
Clone | OTI1H6 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-168-0322
![]() ![]() |
OriGene
TA804606 |
100 μL |
Each of 1 for $516.29
|
|
|||||
Description
ERBB4 Monoclonal antibody specifically detects ERBB4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ERBB4 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Primary |
OTI1H6 | |
Monoclonal | |
Liquid | |
RUO | |
IgG2b | |
-20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title