Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Antibody (8F1), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB500704C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500704C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500704C Supplier Novus Biologicals Supplier No. NB500704C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERCC1 Monoclonal specifically detects ERCC1 in Human, Rat, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ERCC1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8F1
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20
Gene Symbols ERCC1
Host Species Mouse
Immunogen Full length recombinant human ERCC1. [UniProt# P07992]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2067
Target Species Human, Rat, Chinese Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.