Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Antibody (ERCC1/2683), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB378346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378346 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378346 Supplier Novus Biologicals Supplier No. NBP308904C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERCC1 Monoclonal antibody specifically detects ERCC1 in Human samples. It is validated for Western Blot, ELISA, Protein Array

Specifications

Antigen ERCC1
Applications Western Blot, ELISA, Protein Array
Classification Monoclonal
Clone ERCC1/2683
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20
Host Species Mouse
Immunogen Recombinant fragment (around aa 191-281) of human ERCC1 protein (exact sequence is proprietary) (Uniprot: P07992)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2067
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.