Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Antibody (OTI1A3), PerCP, Novus Biologicals™
SDP

Catalog No. NB270662PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270662PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270662PCP Supplier Novus Biologicals Supplier No. NBP270662PCP

Mouse Monoclonal Antibody

ERCC1 Monoclonal specifically detects ERCC1 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen ERCC1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1A3
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20
Gene Symbols ERCC1
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human ERCC1 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2067.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.