Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140803
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0127 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0127 Supplier Bethyl Laboratories, Inc Supplier No. A301337AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan

The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne′s syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.

Specifications

Antigen ERCC3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ERCC3
Gene Accession No. P19447, P49135
Gene Alias BTF2, GTF2H, RAD25, TFIIH, XPB
Gene Symbols ERCC3
Host Species Rabbit
Immunogen Between 732 and 782
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13872, 2071
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.