Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC6L Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB045595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045595 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045595 Supplier Novus Biologicals Supplier No. NBP297609C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERCC6L Polyclonal specifically detects ERCC6L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ERCC6L
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent helicase ERCC6-like, DNA excision repair protein ERCC-6-like, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 6-like, excision repair protein ERCC6-like, FLJ20105, MGC131695, PICHexcision repair cross-complementing rodent repair deficiency complementationgroup 6 - like, PLK1-interacting checkpoint helicase, SNF2/RAD54 family protein, Tumor antigen BJ-HCC-15
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ERCC6L fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54821
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.