Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC8 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB105839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105839 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105839 Supplier Novus Biologicals Supplier No. NBP298792C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERCC8 Polyclonal specifically detects ERCC8 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ERCC8
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ERCC8.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 1161
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.