Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC8 Antibody (OTI2G1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289608 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289608 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289608 Supplier Novus Biologicals Supplier No. NBP270412APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERCC8 Monoclonal antibody specifically detects ERCC8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen ERCC8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G1
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8
Host Species Mouse
Immunogen Full length human recombinant protein of human ERCC8(NP_001007235) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 1161
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.