Learn More
ERF Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | ERF |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2018
![]() |
Bethyl Laboratories, Inc
IHC00392T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2017
![]() |
Bethyl Laboratories, Inc
IHC00392 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Naked mole rat and L
ABT1 (activator of basal transcription 1) is a nuclear protein that associates with the TATA-binding protein (TBP) and enhances basal transcription activity of class II promoters. ABT1 associates with TBP in HeLa nuclear extracts in vitro. Another protein, designated ERF, is a member of the Ets family of transcription factors. The members of the Ets family are grouped because they share a highly conserved DNA binding domain. These factors are involved in growth factor pathways and regulate both proliferation and differentiation. ERF (Ets-2 repressor factor) is a ubiquitously expressed Ets-domain protein that exhibits strong transcriptional repressor activity, suppresses Ets-induced transformation and is regulated by MAPK phosphorylation. ERF transcription may be regulated by Ets-domain proteins. Additionally, modulation of ERF activity is involved in the transcriptional regulation of genes activated during entry into G1 phase.Specifications
ERF | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
PE-2, PE2, ETS domain-containing transcription factor ERF | |
Between 498 and 548 | |
Primary | |
4° C | |
ERF |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P50548, P70459 | |
13875, 2077 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.