Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eRF1 Antibody (4F9H12) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252552 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252552 Supplier Novus Biologicals Supplier No. NBP252552
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

eRF1 Monoclonal specifically detects eRF1 in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Simple Western, Western Blot.

Specifications

Antigen eRF1
Applications CyTOF, ELISA, Flow Cytometry, Simple Western, Western Blot
Classification Monoclonal
Clone 4F9H12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias D5S1995, ERF1eRF1, eukaryotic peptide chain release factor subunit 1, Eukaryotic release factor 1, eukaryotic translation termination factor 1, polypeptide chain release factor 1, Protein Cl1, RF1ERF, sup45 (yeast omnipotent suppressor 45) homolog-like 1, SUP45L1, TB3-1MGC111066
Gene Symbols ETF1
Host Species Mouse
Immunogen Purified recombinant fragment of human RF1 (AA: 288-437) expressed in E. Coli.
Molecular Weight of Antigen 49 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2107
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.