Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eRF1 Antibody (OTI2E3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB270692APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270692APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270692APC Supplier Novus Biologicals Supplier No. NBP270692APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eRF1 Monoclonal specifically detects eRF1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen eRF1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2E3
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias D5S1995, ERF1eRF1, eukaryotic peptide chain release factor subunit 1, Eukaryotic release factor 1, eukaryotic translation termination factor 1, polypeptide chain release factor 1, Protein Cl1, RF1ERF, sup45 (yeast omnipotent suppressor 45) homolog-like 1, SUP45L1, TB3-1MGC111066
Gene Symbols ETF1
Host Species Mouse
Immunogen Full length recombinant protein of human eRF1 (NP_004721) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2107.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.