Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ ProFlex™ 650 Neoprene Elbow Sleeve
Shop All Ergodyne Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
For Use With (Application):
Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis
Proper lifting protection, support without muscle restriction, helps soothe painful strains
Material:
Neoprene
Design:
Contoured
Description
- Provide warmth to covered area
- Soothe painful strains
- Support without muscle restriction
- Conform to the body's natural shape
- Made of neoprene four way stretch fabric
- Single layer
- Ambidextrous
- Color: black
Specifications
Specifications
| Color | Black |
| Closure Type | Adjustable cinch strap |
| Product Type | Elbow Sleeve |
| Design | Contoured |
| Size | Small |
| For Use With (Application) | Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis |
| Hand | Ambidextrous |
| Material | Neoprene |
| Model | 655 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction