Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ Ultra Work Sport Anti-Fatigue Insoles
Shop All Impacto Protective Products Products
Click to view available options
Size:
Men: 11 to 12
Men: 4 to 5; Women: 6 to 7
Men: 5 to 6; Women: 7.5 to 8.5
Men: 6.5 to 7.5; Women: 9 to 10
Men: 8 to 9; Women: 10.5 to 11.5
Men: 9.5 to 10.5; Women: 11.5 to 12
Description
Impacto™ Ultra Work-Sport Anti-Fatigue Insoles
- Sorbothane™ inlays absorb shock and help relieve pain in feet, knees and lower back
- Designed to provide cushioned support and shock absorbing comfort
- Cupped heel for rear foot stability
- Breathable and antifungal
- For use with closed shoes and boots
Specifications
Specifications
Color | Blue |
Length (English) | 12 in. |
Length (Metric) | 30.48 cm |
Material | Sorbothane |
Size | Men: 11 to 12 |
Gender | Unisex |
Product Type | Ergotech Ultra Work Sport Insoles |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction