Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERH Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB350503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350503 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350503 Supplier Novus Biologicals Supplier No. NBP318856MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERH Polyclonal antibody specifically detects ERH in Human samples. It is validated for ELISA

Specifications

Antigen ERH
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias DROER, enhancer of rudimentary (Drosophila) homolog, enhancer of rudimentary homolog, enhancer of rudimentary homolog (Drosophila), FLJ27340
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ERH (Accession#: P84090; Met1-Lys104)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2079
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.