Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK1/ERK2 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB306068 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306068 100 μL
NB306067 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB306068 Supplier Novus Biologicals Supplier No. NBP323530100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERK1/ERK2 Polyclonal antibody specifically detects ERK1/ERK2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen ERK1/ERK2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:50-1:200
Formulation 0.2 um filtered solution in PBS
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to residues around Thr202, Tyr204 of Human phospho-ERK1/ERK2
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.