Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK1/ERK2 [p Tyr187, p Tyr204, p Thr185, p Thr202] Antibody (269434) [Biotin], Novus Biologicals™
SDP

Catalog No. NB402325 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402325 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402325 Supplier Novus Biologicals Supplier No. FAB1018B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen ERK1/ERK2
Applications Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 269434
Conjugate Biotin
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Host Species Rabbit
Immunogen Phosphopeptide containing ERK1 T202/Y204 site
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.