Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK1, p Thr202, p Tyr204 Antibody, Novus Biologicals™
SDP

Catalog No. NB500141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500141 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500141 Supplier Novus Biologicals Supplier No. NB500141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

ERK1 Polyclonal specifically detects ERK1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ERK1 (p Thr202, p Tyr204)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P63086
Gene Alias EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24
Gene Symbols MAPK3
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding ERK1 conjugated to KLH. Accession # P63086
Molecular Weight of Antigen 42 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Breast Cancer, Cancer, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5595
Test Specificity Specific for the ∼42k - 44k ERK/MAPK phosphorylated at Thr202 and Tyr204. Immunolabeling is blocked by the phosphopeptide used as antigen but not by the corresponding dephosphopeptide. The immunolabeling is completely eliminated by lambda-phosphatase.
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.