Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK2 Antibody (007), DyLight 650
SDP

Catalog No. NB087819 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087819 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087819 Supplier Novus Biologicals Supplier No. NBP289215C

Rabbit Monoclonal Antibody

ERK2 Monoclonal specifically detects ERK2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ERK2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 7
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ERK2 (NP_002736.3; Met1-Ser360).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5594
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.