Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK2 Antibody (1A4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP174025R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP174025R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP174025R Supplier Novus Biologicals Supplier No. NBP174025R

Mouse Monoclonal Antibody

ERK2 Monoclonal specifically detects ERK2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ERK2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1A4
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
Gene Symbols MAPK1
Host Species Mouse
Immunogen Recombinant human MAPK1 (1-360) purified from E.coli
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5594
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.