Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK3/MAPK6 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB057238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057238 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057238 Supplier Novus Biologicals Supplier No. NBP299003FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERK3/MAPK6 Polyclonal specifically detects ERK3/MAPK6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ERK3/MAPK6
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp686F03189, EC 2.7.11, EC 2.7.11.24, ERK-3, ERK3p97-MAPK, Extracellular signal-regulated kinase 3, extracellular signal-regulated kinase, p97, HsT17250, MAP kinase 6, MAP kinase isoform p97, MAPK 6, mitogen-activated protein kinase 6, p97MAPK, PRKM6, protein kinase, mitogen-activated 5, protein kinase, mitogen-activated 6
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ERK3/MAPK6.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5597
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.