Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK3/MAPK6 Antibody (OTI5E1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271242 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271242 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271242 Supplier Novus Biologicals Supplier No. NBP271242
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERK3/MAPK6 Monoclonal specifically detects ERK3/MAPK6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ERK3/MAPK6
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5E1
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias DKFZp686F03189, EC 2.7.11, EC 2.7.11.24, ERK-3, ERK3p97-MAPK, Extracellular signal-regulated kinase 3, extracellular signal-regulated kinase, p97, HsT17250, MAP kinase 6, MAP kinase isoform p97, MAPK 6, mitogen-activated protein kinase 6, p97MAPK, PRKM6, protein kinase, mitogen-activated 5, protein kinase, mitogen-activated 6
Gene Symbols MAPK6
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 345-721 of human ERK3
Molecular Weight of Antigen 82.5 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5597
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.