Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK5/BMK1 Antibody (OTI3H2), HRP, Novus Biologicals™
SDP

Catalog No. NBP271243H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271243H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271243H Supplier Novus Biologicals Supplier No. NBP271243H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERK5/BMK1 Monoclonal specifically detects ERK5/BMK1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ERK5/BMK1
Applications Western Blot
Classification Monoclonal
Clone OTI3H2
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias Big MAP kinase 1, BMK-1, BMK1 kinase, BMK1EC 2.7.11.24, EC 2.7.11, ERK-5, ERK5MAPK 7, Extracellular signal-regulated kinase 5, extracellular-signal-regulated kinase 5, MAP kinase 7, mitogen-activated protein kinase 7, PRKM7ERK4
Gene Symbols MAPK7
Host Species Mouse
Immunogen Full length recombinant protein of human ERK5/BMK1 (NP_002740) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5598.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.