Learn More
Description
Specifications
Specifications
| Antigen | ERManI |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 3C2 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | alpha 1,2-mannosidase, EC 3.2.1.113, endoplasmic reticulum alpha-mannosidase 1, endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase, endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1, ER alpha 1,2-mannosidase, ER alpha-1,2-mannosidase, ER ManI, ER mannosidase 1, ERMan1, Man9GlcNAc2-specific processing alpha-mannosidase, Man9GlcNAc2-specific-processing alpha-mannosidase, MANA-ER, Mannosidase alpha class 1B member 1, mannosidase, alpha, class 1B, member 1, MRT15, RP11-350O14.2 |
| Gene Symbols | MAN1B1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
