Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERManI Antibody (3C2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP213167W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213167W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213167W Supplier Novus Biologicals Supplier No. NBP213167AF405

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

ERManI Monoclonal specifically detects ERManI in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen ERManI
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3C2
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. Q9UKM7
Gene Alias alpha 1,2-mannosidase, EC 3.2.1.113, endoplasmic reticulum alpha-mannosidase 1, endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase, endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1, ER alpha 1,2-mannosidase, ER alpha-1,2-mannosidase, ER ManI, ER mannosidase 1, ERMan1, Man9GlcNAc2-specific processing alpha-mannosidase, Man9GlcNAc2-specific-processing alpha-mannosidase, MANA-ER, Mannosidase alpha class 1B member 1, mannosidase, alpha, class 1B, member 1, MRT15, RP11-350O14.2
Gene Symbols MAN1B1
Host Species Mouse
Immunogen Full length human recombinant ERManI [Swiss-Prot# Q9UKM7]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 11253
Test Specificity Human. Does not cross react with Mouse.
Target Species Human, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.